Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

376205

Sigma-Aldrich

Triphenylsilylamine

97%

Synonym(s):

Aminotriphenylsilane, TPSA, Triphenylaminosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3SiNH2
CAS Number:
Molecular Weight:
275.42
Beilstein/REAXYS Number:
2941367
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

mp

60-62 °C (lit.)

storage temp.

2-8°C

SMILES string

N[Si](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C18H17NSi/c19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H,19H2

InChI key

GLQOALGKMKUSBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triphenylsilylamine is a silyl compound.[1]

application

Triphenylsilylamine may be used in the following studies:
  • Preparation of dimers of the type [Ti(μ-NR)(NMe2)2]2 (R = 1-adamantyl, (t)Bu, SiPh3).[2]
  • Silylation during the pretreatment of fused silica capillary surface to achieve a homogeneous film.[3]
  • Preparation of triphenylsilylsulfinylamine, via reaction with thionylchloride.[4]
  • To investigate the molecular tuning effect of silanation treatment for HZSM-5 crystals.[1]
  • Synthesis of 9-aminopyrido[3′,3′:4,5]pyrrolo[1,2-c]pyrimidine (9-aminosubstituted derivative).[5]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cătălin Buiu et al.
Current computer-aided drug design, 10(3), 237-249 (2014-01-01)
Xanthine-based molecules such as serine protease dipeptidyl peptidase 4 (DPP4) inhibitors are compounds often used in improving glycemic control in type 2 diabetic patients and also used for their effects as mild stimulants and as bronchodilators, notably in treating asthma
Gilles H Goetz et al.
Molecular pharmaceutics, 14(2), 386-393 (2016-12-31)
EPSA is an experimental descriptor of molecular polarity obtained from chromatographic retention in supercritical fluid chromatography (SFC) systems, previously shown by Goetz et al. to correlate with passive permeability of cyclic peptides. The present study focuses on EPSA in relation
Meng-Yao Wang et al.
Journal of agricultural and food chemistry, 67(7), 1823-1830 (2019-01-25)
The increasing prevalence of fungal diseases, continual development of resistance, and stringent environmental regulations have revealed an urgent need to develop more selective, safer, resistance-breaking, and cost-effective fungicides. However, most new fungicidal lead compounds fail in their late stages of
Manish Sud
Journal of chemical information and modeling, 56(12), 2292-2297 (2016-12-28)
MayaChemTools is a growing collection of Perl scripts, modules, and classes to support a variety of computational drug discovery needs, such as manipulation and analysis of data, generation of two-dimensional (2D) fingerprints, similarity searching, and calculation of physicochemical properties. MayaChemTools
Bernhard X Mayer-Helm et al.
Journal of separation science, 27(4), 335-342 (2004-09-01)
Two highly phenylated tetramethyl-p-silphenylene-diphenylsiloxane copolymers were coated on fused silica capillary columns and used as stationary phases in GC. The copolymers offered new insights into the coating process and column preparation due to their physicochemical properties. The fused silica capillary

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service