Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

376175

Sigma-Aldrich

1-(Trimethylsilyl)-1-pentyne

98%

Synonym(s):

1-Pentyn-1-yltrimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C≡CSi(CH3)3
CAS Number:
Molecular Weight:
140.30
Beilstein/REAXYS Number:
1744218
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

assay

98%

form

liquid

refractive index

n20/D 1.425 (lit.)

bp

135 °C (lit.)

density

0.765 g/mL at 25 °C (lit.)

SMILES string

CCCC#C[Si](C)(C)C

InChI

1S/C8H16Si/c1-5-6-7-8-9(2,3)4/h5-6H2,1-4H3

InChI key

CABCDUQQPBAHEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-(Trimethylsilyl)-1-pentyne may be used in the synthesis of the natural products credneramide A and B, via Ruthenium-catalyzed Alder-ene reaction.[1] It may be used in the synthesis of 3,3′-ethynediyl-bis-pyridine.[2]

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

86.0 °F - closed cup

flash_point_c

30 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Florian Erver et al.
The Journal of organic chemistry, 77(11), 5215-5219 (2012-05-10)
The first synthesis of the natural products credneramide A and B was accomplished by utilizing Alder-ene reactions between a terminal alkene and an internal alkyne to generate the rather uncommon 1,4-diene substructure of these compounds. Moreover, two different short linear
Copper-free palladium-catalyzed sonogashira-type coupling of aryl halides and 1-aryl-2-(trimethylsilyl) acetylenes.
S?rensen US and Pombo-Villar E.
Tetrahedron, 61(10), 2697-2703 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service