Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

375977

Sigma-Aldrich

Methoxy(dimethyl)octylsilane

98%

Synonym(s):

Dimethylmethoxy-n -octylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7Si(CH3)2OCH3
CAS Number:
Molecular Weight:
202.41
Beilstein/REAXYS Number:
4362666
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

98%

form

liquid

refractive index

n20/D 1.423 (lit.)

bp

221-223 °C (lit.)

density

0.813 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC[Si](C)(C)OC

InChI

1S/C11H26OSi/c1-5-6-7-8-9-10-11-13(3,4)12-2/h5-11H2,1-4H3

InChI key

BAXHQTUUOKMMGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methoxy(dimethyl)octylsilane was used to functionalize partially oxidized porous silicon with silane groups.{89}

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

181.4 °F - closed cup

flash_point_c

83 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huiyuan Hou et al.
Acta biomaterialia, 24, 309-321 (2015-06-19)
Many chorioretinal diseases are chronic and need sustained drug delivery systems to keep therapeutic drug level at the disease site. Many intravitreal drug delivery systems under developing do not have mechanism incorporated for a non-invasive monitoring of drug release. The
Alejandra Nieto et al.
Investigative ophthalmology & visual science, 56(2), 1070-1080 (2015-01-24)
To understand the relationship between rapamycin loading/release and surface chemistries of porous silicon (pSi) to optimize pSi-based intravitreal delivery system. Three types of surface chemical modifications were studied: (1) pSi-COOH, containing 10-carbon aliphatic chains with terminal carboxyl groups grafted via

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service