Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

373923

Sigma-Aldrich

Glycerol propoxylate

average Mn ~1,500

Synonym(s):

Glycerol propoxylate (1PO/OH)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(C3H6O)nCH[CH2(OC3H6)nOH]2
CAS Number:
MDL number:
UNSPSC Code:
12162002

vapor density

>1 (vs air)

vapor pressure

<0.3 mmHg ( 20 °C)

description

functionality 3.0

mol wt

average Mn ~1,500

contains

130-190 ppm proprietary phenolic antioxidant

solubility

H2O: soluble 1.000 g/L

density

1.018 g/mL at 25 °C

InChI

1S/C12H26O9/c13-5-9(16)1-3-19-12(21-8-11(18)7-15)20-4-2-10(17)6-14/h9-18H,1-8H2

InChI key

ZSKSVMDBXFSLMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service