Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

373591

Sigma-Aldrich

2-Methoxyphenethylamine

technical grade, 92%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
151.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

technical grade

assay

92%

bp

236-237 °C (lit.)

density

1.033 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1CCN

InChI

1S/C9H13NO/c1-11-9-5-3-2-4-8(9)6-7-10/h2-5H,6-7,10H2,1H3

InChI key

WSWPCNMLEVZGSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service