Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

372943

Sigma-Aldrich

Chlorotriethylsilane solution

1.0 M in THF

Synonym(s):

TESCl, Triethylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3SiCl
CAS Number:
Molecular Weight:
150.72
Beilstein/REAXYS Number:
1732860
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

liquid

concentration

1.0 M in THF

density

0.886 g/mL at 25 °C

SMILES string

CC[Si](Cl)(CC)CC

InChI

1S/C6H15ClSi/c1-4-8(7,5-2)6-3/h4-6H2,1-3H3

Inchi Key

DCFKHNIGBAHNSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

target_organs

Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

1.4 °F - closed cup

flash_point_c

-17 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service