Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

371297

Sigma-Aldrich

4′-Bromo-(1,1′-biphenyl)-4-ol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4C6H4OH
CAS Number:
Molecular Weight:
249.10
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

assay

97%

form

powder, crystals or chunks

mp

164-166 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccc(cc1)-c2ccc(Br)cc2

InChI

1S/C12H9BrO/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8,14H

InChI key

ARUBXNBYMCVENE-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service