Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

371246

Labclear gas filter

Disposable, weight 200 g(volume)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.51

L × D

7 in. × 2 in.

weight

200 g(volume)

Looking for similar products? Visit Product Comparison Guide

General description

Removes water, oil, and other materials from inert carrier gases such as N2, He, or Ar. Filters contain a mixture of indicating Drierite and 13X molecular sieves. Refillable filter tested at 120 psi up to 100°F, disposable filter tested at 125 psi up to 100°F. Supplied with 1/4 in. compression fittings.

Legal Information

Drierite is a trademark of W.A. Hammond Drierite Co., LTD
Labclear is a trademark of Labclear

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service