370568
1H,1H,2H-Perfluoro-1-octene
99%
Synonym(s):
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octene, 1H,1H,2H-Perfluoro-1-octene
About This Item
Recommended Products
assay
99%
refractive index
n20/D 1.295 (lit.)
bp
102-104 °C (lit.)
density
1.52 g/mL at 25 °C (lit.)
SMILES string
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C=C
InChI
1S/C8H3F13/c1-2-3(9,10)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21/h2H,1H2
InChI key
FYQFWFHDPNXORA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
68.0 °F - closed cup
flash_point_c
20 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service