Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

367486

Sigma-Aldrich

Dimethyl 4-nitrophthalate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3-1,2-(CO2CH3)2
CAS Number:
Molecular Weight:
239.18
Beilstein/REAXYS Number:
1886234
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

bp

192-195 °C/25 mmHg (lit.)

mp

64-66 °C (lit.)

functional group

ester
nitro

SMILES string

COC(=O)c1ccc(cc1C(=O)OC)[N+]([O-])=O

InChI

1S/C10H9NO6/c1-16-9(12)7-4-3-6(11(14)15)5-8(7)10(13)17-2/h3-5H,1-2H3

InChI key

XWBDWELWBUWSNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of highly specific fluorescence immunoassay and enzyme-linked immunosorbent assay for detection of dimethyl phthalate in water samples.
Zhang M, et al.
Food and agricultural immunology, 22(4), 297-309 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service