Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

366714

Sigma-Aldrich

(2R,5S)-5-Methyl-4,4-diphenyl-1,3,2-dioxathiolane

98%

Synonym(s):

(S)-(−)-1,1-Diphenyl-1,2-propanediol cyclic sulfite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O3S
CAS Number:
Molecular Weight:
274.33
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

98%

optical activity

[α]20/D −245°, c = 1 in chloroform

mp

110-113 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H]1OS(=O)OC1(c2ccccc2)c3ccccc3

InChI

1S/C15H14O3S/c1-12-15(18-19(16)17-12,13-8-4-2-5-9-13)14-10-6-3-7-11-14/h2-12H,1H3/t12-,19+/m0/s1

InChI key

ACONWRLGPDTDKX-HXPMCKFVSA-N

Application

Used to synthesize chiral t-butyl sulfoxides.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 30, 3659-3659 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service