Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

366498

Sigma-Aldrich

2-(Methyldiphenylsilyl)ethanol

99%

Synonym(s):

(2-Hydroxyethyl)diphenylmethylsilane, 2-(Diphenylmethylsilyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3Si(C6H5)2CH2CH2OH
CAS Number:
Molecular Weight:
242.39
Beilstein/REAXYS Number:
4428743
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

liquid

refractive index

n20/D 1.580 (lit.)

bp

152 °C/0.2 mmHg (lit.)

density

1.063 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

C[Si](CCO)(c1ccccc1)c2ccccc2

InChI

1S/C15H18OSi/c1-17(13-12-16,14-8-4-2-5-9-14)15-10-6-3-7-11-15/h2-11,16H,12-13H2,1H3

InChI key

RYVPOJDQPLVTLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

2-(Methyldiphenylsilyl)ethanol (2-(Diphenylmethylsilyl)ethanol) may be used in the preparation of 2-diphenylmethylsilylethyldithiophosphorodichloridate.[1]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D C Capaldi et al.
Nucleic acids research, 28(9), E40-E40 (2000-04-11)
A triester method for the synthesis of deoxynucleoside phosphorodithioate dimers is described. The phosphorodithioate linkage is introduced using a new dithiophosphorylating reagent DPSE-SP(S)Cl(2)where DPSE = 2-diphenylmethylsilylethyl. This group is removed quickly using tetra-butylammonium fluoride leading to the quantitative formation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service