Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

365173

Sigma-Aldrich

Tetrakis(decyl)ammonium bromide

99%

Synonym(s):

Tetra-decylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)9]4N(Br)
CAS Number:
Molecular Weight:
659.01
Beilstein/REAXYS Number:
5842649
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Pricing and availability is not currently available.

assay

99%

SMILES string

[Br-].CCCCCCCCCC[N+](CCCCCCCCCC)(CCCCCCCCCC)CCCCCCCCCC

InChI

1S/C40H84N.BrH/c1-5-9-13-17-21-25-29-33-37-41(38-34-30-26-22-18-14-10-6-2,39-35-31-27-23-19-15-11-7-3)40-36-32-28-24-20-16-12-8-4;/h5-40H2,1-4H3;1H/q+1;/p-1

InChI key

AHNISXOXSNAHBZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service