Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

365092

Sigma-Aldrich

Osmium tetroxide on poly(4-vinylpyridine)

Synonym(s):

Osmium tetroxide, ~1 wt. % on poly(4-vinylpyridine), Osmium(VIII) oxide on poly(4-vinylpyridine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OsO4
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: alcohol reactive

storage temp.

2-8°C

SMILES string

O=[Os](=O)(=O)=O

InChI

1S/4O.Os

InChI key

VUVGYHUDAICLFK-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Ox. Sol. 2

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service