Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

364886

Sigma-Aldrich

(R)-(−)-1-Benzyloxy-3-(p-tosyloxy)-2-propanol

95%

Synonym(s):

(R)-(−)-1-Benzyloxy-3-tosyloxy-2-propanol, (R)-(−)-3-Benzyloxy-1,2-propanediol 1-(p-toluenesulfonate), 1-Benzyl-sn-glycerol 3-tosylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO3CH2CH(OH)CH2OCH2C6H5
CAS Number:
Molecular Weight:
336.40
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

95%

optical activity

[α]25/D −7°, c = 10 in toluene

mp

52-55 °C (lit.)

functional group

ether
hydroxyl
phenyl
tosylate

SMILES string

Cc1ccc(cc1)S(=O)(=O)OC[C@H](O)COCc2ccccc2

InChI

1S/C17H20O5S/c1-14-7-9-17(10-8-14)23(19,20)22-13-16(18)12-21-11-15-5-3-2-4-6-15/h2-10,16,18H,11-13H2,1H3/t16-/m1/s1

InChI key

HBMUWOSOGHUWSS-MRXNPFEDSA-N

Application

Reagent employed in enantioselective carbohydrate, terpene, and alkaloid syntheses.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 42, 447-447 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service