Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

364282

Sigma-Aldrich

Di-μ-chlorobis[2-[(dimethylamino)methyl]phenyl-C,N]dipalladium(II)

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$143.00

$143.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$143.00

About This Item

Empirical Formula (Hill Notation):
C18H24Cl2N2Pd2
CAS Number:
Molecular Weight:
552.14
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$143.00


Check Cart for Availability

Request a Bulk Order

assay

98%

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

175 °C (dec.) (lit.)

SMILES string

CN(C)Cc1ccccc1[Pd]Cl.CN(C)Cc2ccccc2[Pd]Cl

InChI

1S/2C9H12N.2ClH.2Pd/c2*1-10(2)8-9-6-4-3-5-7-9;;;;/h2*3-6H,8H2,1-2H3;2*1H;;/q;;;;2*+1/p-2

InChI key

DSRDIGYUGAOPJI-UHFFFAOYSA-L

application

Catalyst for:
  • Oxidative allylic C-H silylation
  • Transesterification reactions
  • Suzuki-Miyaura cross-coupling reactions
  • Stereocontrolled homologation of carbon chain boronic esters using Hoppe′s lithiated carbamates
  • Oxidative carbonylation reactions
  • Buchwald-Hartwig reactions
  • Mizoroki-Heck reaction

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service