363111
Poly(vinyl alcohol)
average Mw 85,000-124,000, 96% hydrolyzed
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
crystalline powder
crystals or granules
powder
mol wt
average Mw 85,000-124,000
falling ball
25-29 cP, 4 % in H2O(20 °C)(lit.)
InChI
1S/C2H4O/c1-2-3/h2-3H,1H2
InChI key
IMROMDMJAWUWLK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
No data available
flash_point_c
No data available
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
XiaoJun Zuo et al.
Carbohydrate polymers, 92(2), 2181-2186 (2013-02-13)
Novel poly(vinyl alcohol)/citric acid/chitosan (PVA/CA/CHT, PCC) beads were prepared as an adsorbent for the removal of trivalent chromium (Cr(3+)) in aqueous solutions. PCC beads with different mass ratios (I, II, III, IV, V, ad VI) between PVA/CA/CHT were evaluated for
Selby Maphutha et al.
Scientific reports, 3, 1509-1509 (2013-03-23)
A carbon nanotube (CNT) integrated polymer composite membrane with a polyvinyl alcohol barrier layer has been prepared to separate oil from water for treatment of oil-containing waste water. The CNTs were synthesised using chemical vapour deposition, and a phase inversion
Gang Wang et al.
PloS one, 6(11), e27605-e27605 (2011-11-24)
Mucosal vaccination has been demonstrated to be an effective means of eliciting protective immunity against aerosol infections of foot and mouth disease virus (FMDV) and various approaches have been used to improve mucosal response to this pathogen. In this study
Wei Li et al.
Carbohydrate polymers, 92(2), 2232-2238 (2013-02-13)
Poly(vinyl alcohol) (PVA)/sodium alginate (ALG)/organic rectorite (OREC) composite nanofibrous mats are fabricated by electrospinning aqueous solutions with different mixing ratios. Both good fiber shape and three-dimensional structure of nanofibrous mats can be observed by Field Emission Scanning Electron Microscopy. Energy-dispersive
Hye Rim Cho et al.
Anticancer research, 33(1), 223-230 (2012-12-26)
To evaluate the feasibility of the McA-RH7777 tumor model in Sprague-Dawley (SD) rats, for study of hepatoma and transarterial chemoembolization. McA-RH7777 rat hepatoma cells (1×10(7)) were inoculated into the left hepatic lobe of SD rats (n=38). Chemoembolization with left common
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service