Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

362239

Sigma-Aldrich

cis-1,2-Cyclooctanediol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H14(OH)2
CAS Number:
Molecular Weight:
144.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

form

solid

mp

78-80 °C (lit.)

SMILES string

O[C@H]1CCCCCC[C@H]1O

InChI

1S/C8H16O2/c9-7-5-3-1-2-4-6-8(7)10/h7-10H,1-6H2/t7-,8+

InChI key

HUSOFJYAGDTKSK-OCAPTIKFSA-N

General description

cis-1,2-Cyclooctanediol is a 1,2-disubstituted acyclic ethylene glycol.[1] It undergoes cyclocondensation with oxalyl chloride in the presence of triethylamine at 0°C to yield cyclic oxalate.[1]

application

cis-1,2-Cyclooctanediol may be used in the preparation of suberic acid.[2]

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes W de Boer et al.
Dalton transactions (Cambridge, England : 2003), (44)(44), 6283-6295 (2008-11-06)
The role played by the additives salicylic acid, L-ascorbic acid and oxalic acid in promoting the catalytic activity of [MnIV2(O)3(tmtacn)2](PF6)2 (1(PF6)2, where tmtacn = N,N',N''-trimethyl-1,4,7-triazacyclononane) in the epoxidation and cis-dihydroxylation of alkenes with H2O2 and in suppressing the catalysed decomposition
Taisuke Itaya et al.
Chemical & pharmaceutical bulletin, 50(1), 83-86 (2002-02-05)
The relationship between the product patterns and the configurations of 1,2-cycloheptane- and 1,2-cyclooctanediols 9 in the cyclocondensations with oxalyl chloride in the presence of triethylamine at 0 degrees C has been shown analogous to that obtained for 1,2-disubstituted acyclic ethylene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service