359882
4-(Dimethylamino)pyridine, polymer-bound
200-400 mesh, extent of labeling: ~6.0 mmol/g loading, 2 % cross-linked
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H8
CAS Number:
Molecular Weight:
104.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
powder
crosslinking
2 % cross-linked
reaction suitability
reaction type: solution phase peptide synthesis
extent of labeling
~6.0 mmol/g loading
particle size
200-400 mesh
Looking for similar products? Visit Product Comparison Guide
Application
Polymer-bound equivalent of DMAP. Can be used as a catalyst for acylation reactions.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
S. Shinkai et al.
Bulletin of the Chemical Society of Japan, 54, 631-631 (1981)
M. Tomoi et al.
Macromol. Chem. Rapid Commun., 3, 537-537 (1982)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service