358916
Vinylene trithiocarbonate
98%
Synonym(s):
1,3-Dithiole-2-thione
About This Item
Recommended Products
assay
98%
form
crystals
mp
48-50 °C (lit.)
functional group
thioether
SMILES string
S=C1SC=CS1
InChI
1S/C3H2S3/c4-3-5-1-2-6-3/h1-2H
InChI key
WYKJWNVWJOKVQP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service