Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

357154

Sigma-Aldrich

Vanadium

foil, thickness 1.0 mm, 99.7%

Synonym(s):

Atomic vanadium, Vanadium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
V
CAS Number:
Molecular Weight:
50.94
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor pressure

8 mmHg ( 20 °C)

assay

99.7%

form

foil

resistivity

24.8-26.0 μΩ-cm, 20°C

thickness

1.0 mm

bp

3380 °C (lit.)

mp

1890 °C (lit.)

density

6.11 g/mL at 25 °C (lit.)

SMILES string

[V]

InChI

1S/V

InChI key

LEONUFNNVUYDNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

3.8 g = 25 × 25 mm

comparable product

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biswajit Mukherjee et al.
Toxicology letters, 150(2), 135-143 (2004-04-20)
The biological image of the transition element vanadium ferments a great deal of contradiction-from toxicity to essentiality. Importance of this element as micro-nutrient is yet to be unequivocally accepted by biologists and biomedical scientists. In spite of toxicity, it seems
Shyam Biswas et al.
Physical chemistry chemical physics : PCCP, 15(10), 3552-3561 (2013-02-06)
Two perfluorinated metal hydroxo terephthalates [M(III)(OH)(BDC-F)]·n(guests) (M(III) = V, MIL-47-F-AS or 1-AS; Al, Al-MIL-53-F-AS or 2-AS) (BDC-F = 2-fluoro-1,4-benzenedicarboxylate; AS = as-synthesized) have been synthesized by a hydrothermal method using microwave irradiation (1-AS) or conventional electric heating (2-AS), respectively. The
Nolan S Horner et al.
Analytica chimica acta, 758, 28-35 (2012-12-19)
A previously developed method based on continuous on-line leaching with artificial gastro-intestinal fluids was used to determine the bio-accessible fraction of As, Cu, Fe, V and Zn in brown and white rice from California by inductively coupled mass spectrometry (ICP-MS).
A V Kudrin
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS), 14(3), 129-142 (2000-12-29)
In recent years several studies have shown that NF-kappaB might be a very important therapeutic target in the treatment ot various chronic inflammatory, degenerative and tumour diseases. Trace elements play essential roles in the regulation ot cell signaling mechanisms via
D G Barceloux
Journal of toxicology. Clinical toxicology, 37(2), 265-278 (1999-06-26)
Vanadium is a steel-grey, corrosion-resistant metal, which exists in oxidation states ranging from -1 to +5. Metallic vanadium does not occur in nature, and the most common valence states are +3, +4, and +5. The pentavalent form (VO3-) predominates in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service