35650
1,2-Dichloroethyl ethyl ether
technical, ~85% (GC)
Synonym(s):
1,2-Dichloro-1-ethoxyethane
About This Item
Recommended Products
grade
technical
assay
~85% (GC)
refractive index
n20/D 1.442 (lit.)
n20/D 1.442
bp
140-145 °C (lit.)
density
1.167 g/mL at 25 °C (lit.)
SMILES string
CCOC(Cl)CCl
InChI
1S/C4H8Cl2O/c1-2-7-4(6)3-5/h4H,2-3H2,1H3
InChI key
NNBUKAPOVBEMNI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
107.6 °F - closed cup
flash_point_c
42 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service