About This Item
Recommended Products
assay
95%
mp
411-412 °C (lit.)
SMILES string
CC(C)(C)c1cc2Cc3cc(cc(Cc4cc(cc(Cc5cc(cc(Cc6cc(cc(Cc7cc(cc(Cc8cc(cc(Cc9cc(cc(Cc(c1)c2O)c9O)C(C)(C)C)c8O)C(C)(C)C)c7O)C(C)(C)C)c6O)C(C)(C)C)c5O)C(C)(C)C)c4O)C(C)(C)C)c3O)C(C)(C)C
InChI
1S/C88H112O8/c1-81(2,3)65-33-49-25-51-35-66(82(4,5)6)37-53(74(51)90)27-55-39-68(84(10,11)12)41-57(76(55)92)29-59-43-70(86(16,17)18)45-61(78(59)94)31-63-47-72(88(22,23)24)48-64(80(63)96)32-62-46-71(87(19,20)21)44-60(79(62)95)30-58-42-69(85(13,14)15)40-56(77(58)93)28-54-38-67(83(7,8)9)36-52(75(54)91)26-50(34-65)73(49)89/h33-48,89-96H,25-32H2,1-24H3
Inchi Key
OLZFZIXORGGLLS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service