Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34655

Sigma-Aldrich

Di-tert-butylcyclopentadiene

mixture of isomers, ≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22
CAS Number:
Molecular Weight:
178.31
Beilstein/REAXYS Number:
3537135
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.467

density

0.836 g/mL at 20 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

CC(C)(C)C1C=CC(=C1)C(C)(C)C

InChI

1S/C13H22/c1-12(2,3)10-7-8-11(9-10)13(4,5)6/h7-10H,1-6H3

InChI key

RADVPPBTEASJRZ-UHFFFAOYSA-N

General description

Preparation of di-tert-butylcyclopentadiene by phase-transfer-catalyzed alkylation has been reported.[1] It reacts with excess potassium tert-butoxide in the presence of elemental selenium to yield cyclopentadienes bridged by two or three selenium atoms through one or two diselenoketal functionalities.[2] It reacts with excess potassium tert-butoxide in the presence of elemental sulfur to yield cyclopentadienes bridged by sulfur atoms via one or two dithioketal functionalities.[3]

Application

Di-tert-butylcyclopentadiene may be used in the preparation of 2,5,4′,6′-tetra-tert-butyl-5′H-spirocyclopentane-1,2′-cyclopenta[b][1,3]diselena-2,4-diene and 1,3,5,7-tetra-tert-butyl-3a,7a-dihydro-3a,7a-epi-selenodicyclopenta[b,e][1,4]diselenine.[2]

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

48.2 °F - closed cup

flash_point_c

9 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclopentadienone Cyclopentadienediyl Diselenoketals.
Thaler G, et al.
European Journal of Inorganic Chemistry, 7, 973-976 (1998)
Di-tert-butylcyclopentadiene and tri-tert-butylcyclopentadiene.
Venier CG and Casserly EW.
Journal of the American Chemical Society, 112(7), 2808-2809 (1990)
Cyclopentadienone cyclopentadienediyl dithioketals.
Thaler G, et al.
Inorgorganica Chimica Acta, 304(2), 184-189 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service