Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

345881

Sigma-Aldrich

Zinc 3,5-di-tert-butylsalicylate

97%

Synonym(s):

3,5-Bis(1,1-dimethylethyl)-2-hydroxybenzoic acid zinc(II) complex, 3,5-Di-tert-butylsalicylic acid zinc salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[(CH3)3C]2C6H2-2-(OH)CO2]2Zn
CAS Number:
Molecular Weight:
564.04
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

97%

reaction suitability

reagent type: catalyst
core: zinc

mp

256 °C (dec.) (lit.)

SMILES string

CC(C)(C)c1cc(C(=O)O[Zn]OC(=O)c2cc(cc(c2O)C(C)(C)C)C(C)(C)C)c(O)c(c1)C(C)(C)C

InChI

1S/2C15H22O3.Zn/c2*1-14(2,3)9-7-10(13(17)18)12(16)11(8-9)15(4,5)6;/h2*7-8,16H,1-6H3,(H,17,18);/q;;+2/p-2

InChI key

HCOFMIWUFBMIPV-UHFFFAOYSA-L

Application

Study of reactivity toward Folin-Ciocalteu reagent

Modifier for application of fluorescence to elucidate ground state electronic structure of boron, zinc, and aluminum complexes

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service