Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

344583

Sigma-Aldrich

Chloromethyl trifluoromethyl sulfide

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SCH2Cl
CAS Number:
Molecular Weight:
150.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

assay

90%

form

liquid

refractive index

n20/D 1.3818 (lit.)

bp

63.5 °C/740 mmHg (lit.)

density

1.412 g/mL at 25 °C (lit.)

functional group

chloro
fluoro
thioether

storage temp.

2-8°C

SMILES string

FC(F)(F)SCCl

InChI

1S/C2H2ClF3S/c3-1-7-2(4,5)6/h1H2

InChI key

CQQRPMWQQUKPMR-UHFFFAOYSA-N

Application

Chloromethyl trifluoromethyl sulfide may be used in chemical synthesis.

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

33.8 °F - closed cup

flash_point_c

1 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service