Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

341770

Sigma-Aldrich

o-Toluenesulfonyl isocyanate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NCO
CAS Number:
Molecular Weight:
197.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

refractive index

n20/D 1.54 (lit.)

density

1.303 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1ccccc1S(=O)(=O)N=C=O

InChI

1S/C8H7NO3S/c1-7-4-2-3-5-8(7)13(11,12)9-6-10/h2-5H,1H3

InChI key

HEBTZZBBPUFAFE-UHFFFAOYSA-N

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

supp_hazards

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F R van de Voort et al.
Analytica chimica acta, 918, 1-7 (2016-04-06)
A simple and accurate method for the determination of moisture in edible oils by differential FTIR spectroscopy has been devised based on the stoichiometric reaction of the moisture in oil with toluenesulfonyl isocyanate (TSI) to produce CO2. Calibration standards were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service