Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

340383

Sigma-Aldrich

1-Amino-1-deoxy-D-sorbitol

98%

Synonym(s):

1-Amino-1-deoxysorbitol, D-Glucamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15NO5
CAS Number:
Molecular Weight:
181.19
Beilstein/REAXYS Number:
1721895
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

98%

optical activity

[α]20/D −7.0°, neat

autoignition temp.

~734 °F

mp

127 °C (dec.) (lit.)

SMILES string

NC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H15NO5/c7-1-3(9)5(11)6(12)4(10)2-8/h3-6,8-12H,1-2,7H2/t3-,4+,5+,6+/m0/s1

InChI key

SDOFMBGMRVAJNF-SLPGGIOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi Qin et al.
CNS neuroscience & therapeutics, 23(2), 162-173 (2016-12-13)
Mutations in leucine-rich repeat kinase 2 (LRRK2) are the most prevalent cause of familial and sporadic Parkinson's disease (PD). Because most pathogenic LRRK2 mutations result in enhanced kinase activity, it suggests that LRRK2 inhibitors may serve as a potential treatment
Ryuto Hara et al.
Neuroscience letters, 728, 134958-134958 (2020-04-13)
We recently showed that the mesolimbic dopaminergic system was tonically suppressed during chronic pain by enhanced corticotropin releasing factor (CRF) signaling within the dorsolateral bed nucleus of the stria terminalis (dlBNST), and that inhibition of intra-dlBNST CRF signaling restored the
Iurii Semenov et al.
Bioelectrochemistry (Amsterdam, Netherlands), 105, 65-71 (2015-05-27)
We tested if picosecond electric pulses (psEP; 190 kV/cm, 500 ps at 50% height), which are much shorter than channel activation time, can activate voltage-gated (VG) channels. Cytosolic Ca(2+) was monitored by Fura-2 ratiometric imaging in GH3 and NG108 cells
Karima Ait Ouares et al.
The European journal of neuroscience, 49(1), 6-26 (2018-11-06)
Optogenetics is based on the selective expression of exogenous opsins by neurons allowing experimental control of their electrical activity using visible light. The interpretation of the results of optogenetic experiments is based on the assumption that light stimulation selectively acts
William D Hendricks et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(22), 10994-10999 (2019-05-16)
In temporal lobe epilepsy, sprouting of hippocampal mossy fiber axons onto dentate granule cell dendrites creates a recurrent excitatory network. However, unlike mossy fibers projecting to CA3, sprouted mossy fiber synapses depress upon repetitive activation. Thus, despite their proximal location

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service