Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

339954

Sigma-Aldrich

Tetrahydro-3-furoic acid

contains 250 ppm BHT as inhibitor, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O3
CAS Number:
Molecular Weight:
116.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

assay

99%

form

liquid

contains

250 ppm BHT as inhibitor

refractive index

n20/D 1.4605 (lit.)

bp

140 °C/15 mmHg (lit.)

density

1.214 g/mL at 25 °C (lit.)

functional group

carboxylic acid
ether

SMILES string

OC(=O)C1CCOC1

InChI

1S/C5H8O3/c6-5(7)4-1-2-8-3-4/h4H,1-3H2,(H,6,7)

InChI key

BOTREHHXSQGWTR-UHFFFAOYSA-N

Application

Tetrahydro-3-furoic acid was used in synthesis of o-ethylphenyl tetrahydro-3-furanyl ketone.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic Support for the α-Erythroidine Structure. The Conversion of α-to β-Erythrodine1.
Boekelheide V and Morrison GC.
Journal of the American Chemical Society, 80(15), 3905-3908 (1958)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service