Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

339334

Sigma-Aldrich

Molybdenum(III) chloride

99.95% trace metals basis

Synonym(s):

Molybdenum trichloride, Trichloromolybdenum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MoCl3
CAS Number:
Molecular Weight:
202.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

assay

99.95% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: molybdenum

impurities

≤550.0 ppm Trace Metal Analysis

solubility

H2O: insoluble(lit.)

density

3.58 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[Mo](Cl)Cl

InChI

1S/3ClH.Mo/h3*1H;/q;;;+3/p-3

InChI key

ZSSVQAGPXAAOPV-UHFFFAOYSA-K

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kristina Hackeloer et al.
Organic letters, 13(5), 916-919 (2011-02-02)
A domino sequence involving various MoCl(5)-mediated oxidations followed by trapping and supposed [3,3]-sigmatropic rearrangement provides a fast access to the full carbon skeleton of metasequirin-B. A variety of different moieties R(1) and R(2) are tolerated.
Mladen Litvić et al.
Bioorganic & medicinal chemistry letters, 22(11), 3676-3681 (2012-05-02)
Mo(VI) and Mo(V) salts both react selectively with Hantzsch esters to produce substitute pyridines in good-to-excellent yield (75-99%). The remarkable reactivity and selectivity of MoOCl(4) under reflux of acetonitrile and MoCl(5) in dichloromethane at room temperature encouraged us to propose
H I Ogawa et al.
Mutation research, 320(1-2), 133-140 (1994-01-01)
A series of metal chlorides were subjected to the wing spot test of Drosophila melanogaster. In the test, larvae trans-heterozygous for the wing-hair mutations mwh and flr were orally treated at the third instar stage with a test compound and
Simon Trosien et al.
Organic letters, 14(12), 2976-2979 (2012-06-05)
The strong oxidative power of molybdenum pentachloride gives rise to an efficient oxidative C-C bond formation of benzil derivatives to the corresponding 9,10-phenanthrenequinones. A highly complementary method to previous approaches was developed. The required derivatives are accessible in a modular

Articles

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Protocols

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service