339326
Mercury(II) fluoride
97%
Synonym(s):
Mercuric fluoride
About This Item
Recommended Products
assay
97%
form
powder
reaction suitability
reagent type: catalyst
core: mercury
SMILES string
F[Hg]F
InChI
1S/2FH.Hg/h2*1H;/q;;+2/p-2
Inchi Key
FMSYTQMJOCCCQS-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Disclaimer

signalword
Danger
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service