Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

339148

Sigma-Aldrich

Barium isopropoxide

technical grade

Synonym(s):

Barium diisopropoxide, Barium(2+) isopropoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(OCH(CH3)2)2
CAS Number:
Molecular Weight:
255.50
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

technical grade

form

solid

composition

Ba, 49.5-56.1% gravimetric

reaction suitability

core: barium

mp

200 °C (dec.) (lit.)

SMILES string

CC(C)O[Ba]OC(C)C

InChI

1S/2C3H7O.Ba/c2*1-3(2)4;/h2*3H,1-2H3;/q2*-1;+2

InChI key

CPUJSIVIXCTVEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Self-heat. 2

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service