Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

336777

Sigma-Aldrich

Cobalt boride, mixture of 2:1 and 3:1

99%

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352300

assay

99%

form

powder

particle size

−10 mesh

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Aerosol 3 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

2B - Aerosol cans and lighters

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service