Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33537

Sigma-Aldrich

5,6,14,15-Dibenzo-1,4-dioxa-8,12-diazacyclopentadeca-5,14-diene

≥98.0% (NT)

Synonym(s):

6,7,8,9,10,11,17,18-Octahydro-5H-dibenzo[e,n]-1,4,8,12-dioxadiazacyclopentadecene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N2O2
CAS Number:
Molecular Weight:
312.41
Beilstein/REAXYS Number:
1023420
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

≥98.0% (NT)

form

solid

mp

142-144 °C

SMILES string

C1CNCc2ccccc2OCCOc3ccccc3CNC1

InChI

1S/C19H24N2O2/c1-3-8-18-16(6-1)14-20-10-5-11-21-15-17-7-2-4-9-19(17)23-13-12-22-18/h1-4,6-9,20-21H,5,10-15H2

InChI key

CHCYHXIUMOKQRL-UHFFFAOYSA-N

Other Notes

Mixed donor macrocyclic ligand; complexes with transition metals[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L.F. Lindoy et al.
Cation Binding by Macrocycles null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service