Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

335169

Sigma-Aldrich

Lead(II) carbonate

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbCO3
CAS Number:
Molecular Weight:
267.21
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

assay

≥99.99% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: lead

SMILES string

[PbH2++].[O-]C([O-])=O

InChI

1S/CH2O3.Pb/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

MFEVGQHCNVXMER-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isotopic characterization of six major brands of white basic lead carbonate paint pigments.
M B Rabinowitz et al.
Bulletin of environmental contamination and toxicology, 69(5), 617-623 (2002-10-11)
Brian Gulson et al.
Environmental health perspectives, 117(1), 148-154 (2009-01-24)
In late 2006, the seaside community in Esperance, Western Australia, was alerted to thousands of native bird species dying. The source of the lead was thought to derive from the handling of Pb carbonate concentrate from the Magellan mine through
John Weaver et al.
The Journal of pharmacology and experimental therapeutics, 302(2), 781-786 (2002-07-20)
Nitric-oxide synthase (NOS; EC 1.14.13.39) catalyzes the oxidation of L-arginine to nitric oxide (NO(.)) and L-citrulline via the intermediate N(omega)-hydroxy-L-arginine. Of the three distinct isoforms of NOS that have been characterized, the constitutive neuronal NOS (NOS I) generates NO(.) associated
Haizhou Liu et al.
Environmental science & technology, 42(9), 3241-3247 (2008-06-05)
Reactions of representative lead (II) solid phases (hydrocerussite, cerussite) with chlorine were examined in this study. Chlorine consumption profiles for these solids exhibited a lag phase, during which little consumption of chlorine occurred, and an ensuing rapid reaction phase. The
E Ann Ellis
Methods in molecular biology (Clifton, N.J.), 369, 97-106 (2007-07-28)
Poststaining ultrathin sections on grids is an essential process in preparing specimens for examination using transmission electron microscopy. This process commonly consists of staining in aqueous solutions of uranyl acetate followed by lead citrate and can result in good consistent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service