Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

333867

Sigma-Aldrich

Zinc tetrafluoroborate solution

40 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B2F8Zn
CAS Number:
Molecular Weight:
239.00
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

concentration

40 wt. % in H2O

SMILES string

[Zn++].F[B-](F)(F)F.F[B-](F)(F)F

InChI

1S/2BF4.Zn/c2*2-1(3,4)5;/q2*-1;+2

InChI key

BUMIVIYHDVNOQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properites.[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy C. Higgs et al.
Inorganic chemistry, 37(9), 2263-2272 (2001-10-24)
The mixed functionality pyrazole/phenol ligand (2-hydroxyphenyl)bis(pyrazolyl)methane, L1OH, has been used to prepare a series of linear trimetallic systems with the general structural motif [M(3)(L1O)(4)](2+), where M = Mn(2+), Co(2+), Ni(2+), Cu(2+), and Zn(2+). Each of these complexes has been structurally

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service