Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

333689

Sigma-Aldrich

4-Vinylaniline

technical grade, 90%

Synonym(s):

4-Aminostyrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC6H4NH2
CAS Number:
Molecular Weight:
119.16
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

technical grade

assay

90%

refractive index

n20/D 1.626 (lit.)

bp

213-214 °C (lit.)

density

1.017 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Nc1ccc(C=C)cc1

InChI

1S/C8H9N/c1-2-7-3-5-8(9)6-4-7/h2-6H,1,9H2

InChI key

LBSXSAXOLABXMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

210.0 °F - closed cup

flash_point_c

98.9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service