Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

332100

Sigma-Aldrich

Dimethyl adipate

98%

Synonym(s):

DBE 6 dibasic ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCO(CH2)4COOCH3
CAS Number:
Molecular Weight:
174.19
Beilstein/REAXYS Number:
1707443
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor pressure

0.2 mmHg ( 20 °C)

assay

98%

form

liquid

autoignition temp.

680 °F

expl. lim.

8.1 %

refractive index

n20/D 1.428 (lit.)

bp

109-110 °C/14 mmHg (lit.)

mp

8 °C (lit.)

density

1.062 g/mL at 20 °C (lit.)

SMILES string

COC(=O)CCCCC(=O)OC

InChI

1S/C8H14O4/c1-11-7(9)5-3-4-6-8(10)12-2/h3-6H2,1-2H3

InChI key

UDSFAEKRVUSQDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S A Simpson et al.
Archives of toxicology, 79(4), 224-230 (2005-02-04)
We have previously developed a rat nasal explant system for investigating upper respiratory tract toxicity, and the aims of this study were to determine whether heat shock protein (HSP) 70 is induced in this model following exposure to carbon tetrachloride
M A Medinsky et al.
Toxicology, 160(1-3), 165-172 (2001-03-14)
Inhalation is a common route by which individuals are exposed to toxicants. The air contains a multitude of gases and vapors that are brought into the respiratory tract with each breath. Depending upon the physical and chemical characteristics of the
B A Trela et al.
Experimental and molecular pathology, 56(3), 208-218 (1992-06-01)
Dibasic esters (DBE) solvent has been demonstrated to induce a mild degeneration of the olfactory, but not the respiratory epithelium of the rat nasal cavity following a 90-day inhalation exposure. Previous work has demonstrated that acid phosphatase release is a
M S Bogdanffy et al.
Drug metabolism and disposition: the biological fate of chemicals, 19(1), 124-129 (1991-01-01)
Inhalation exposure of rats to dibasic esters revealed lesions of the nasal olfactory epithelium similar to those observed with other ester solvents. Female rats are more sensitive to these effects than are male rats. It has been proposed that carboxylesterase
J B Morris et al.
Toxicology and applied pharmacology, 108(3), 538-546 (1991-05-01)
Inhalation exposure of the male and female rat to high concentrations of a mixture of the dibasic esters dimethyl succinate (DMS), dimethyl glutarate (DMG), and dimethyl adipate (DMA) results in mild olfactory toxicity. This response is thought to be due

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service