Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

331872

Sigma-Aldrich

(2R,3S)-(−)-N-Z-6-oxo-2,3-diphenylmorpholine

98%

Synonym(s):

Benzyl (2R,3S)-(−)-6-oxo-2,3-diphenyl-4-morpholinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21NO4
CAS Number:
Molecular Weight:
387.43
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

optical activity

[α]25/D −66°, c = 5.5 in methylene chloride

mp

205-207 °C (lit.)

functional group

ester
phenyl

storage temp.

2-8°C

SMILES string

O=C1CN([C@H]([C@H](O1)c2ccccc2)c3ccccc3)C(=O)OCc4ccccc4

InChI

1S/C24H21NO4/c26-21-16-25(24(27)28-17-18-10-4-1-5-11-18)22(19-12-6-2-7-13-19)23(29-21)20-14-8-3-9-15-20/h1-15,22-23H,16-17H2/t22-,23+/m0/s1

InChI key

HECRUWTZAMPQOS-XZOQPEGZSA-N

Application

(2R,3S)-(−)-N-Z-6-oxo-2,3-diphenylmorpholine is a Williams chiral auxiliary, which can be used:
  • As a template in the preparation of Fmoc-L-cyclopentylglycine, which acts as an angiotensin II antagonist.[1]
  • In the synthesis of (L)-Fmoc-α-Me-Lys(Boc)-OH, a building block for the synthesis of peptidomimetics.[2]
  • To introduce chirality in the modified amino acid named N-Fmoc 4-(2′-(di-tert-butylmalonyl))-l-phenylalanine.[3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient asymmetric synthesis of Fmoc-L-cyclopentylglycine via diastereoselective alkylation of glycine enolate equivalent
Singh S and Pennington MW
Tetrahedron Letters, 44(13), 2683-2685 (2003)
Enantioselective synthesis of (L)-Fmoc-alpha-Me-Lys (Boc)-OH via diastereoselective alkylation of oxazinone as a chiral auxiliary
Chauhan SS
Tetrahedron Letters, 50(49), 6913-6915 (2009)
Stereoselective preparation of L-4-(2′-malonyl) phenylalanine suitably protected for Fmoc-based synthesis of potent signal transduction inhibitory ligands
Gao Y and Burke Jr TR
Synlett, 2000(01), 134-136 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service