Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

331848

Sigma-Aldrich

(2R,3S)-(−)-N-Boc-6-oxo-2,3-diphenylmorpholine

98%

Synonym(s):

tert-Butyl (2R,3S)-(−)-6-oxo-2,3-diphenyl-4-morpholinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO4
CAS Number:
Molecular Weight:
353.41
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

98%

optical activity

[α]25/D −87°, c = 5.5 in methylene chloride

optical purity

ee: 99% (HPLC)

mp

206 °C (dec.) (lit.)

functional group

ester
phenyl

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CC(=O)O[C@@H]([C@@H]1c2ccccc2)c3ccccc3

InChI

1S/C21H23NO4/c1-21(2,3)26-20(24)22-14-17(23)25-19(16-12-8-5-9-13-16)18(22)15-10-6-4-7-11-15/h4-13,18-19H,14H2,1-3H3/t18-,19+/m0/s1

InChI key

MRUKRSQUUNYOFK-RBUKOAKNSA-N

Application

(2R,3S)-(-)-N-Boc-6-oxo-2,3-diphenylmorpholine (Williams′ morpholinone) can be used as a reactant to prepare:
  • Indole substituted tryptophan derivatives by alkylation with quaternary gramine in the presence of a base.[1]
  • Tyrosine derivatives, which can be utilized as building blocks for the total synthesis of ecteinascidin and safracin family of alkaloids.[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic studies on ecteinascidin 743: asymmetric synthesis of the versatile amino acid component
Jin, Wei and Williams, Robert M
Tetrahedron Letters, 44(25), 4635-4639 (2003)
Synthesis of tryptophans by alkylation of chiral glycine enolate equivalents with quaternary gramines
Reinfelds, M, et al.
Tetrahedron Letters, 56(43), 5882-5885 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service