Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330655

Sigma-Aldrich

4-Chlorotetrahydropyran

96%

Synonym(s):

4-Chloro-tetrahydro-2H-pyran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9ClO
CAS Number:
Molecular Weight:
120.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

refractive index

n20/D 1.462 (lit.)

bp

150 °C (lit.)

solubility

water: soluble(lit.)

density

1.114 g/mL at 25 °C (lit.)

SMILES string

ClC1CCOCC1

InChI

1S/C5H9ClO/c6-5-1-3-7-4-2-5/h5H,1-4H2

InChI key

DHRSKOBIDIDMJZ-UHFFFAOYSA-N

General description

4-Chlorotetrahydropyran is a commonly employed starting material in organic synthesis.[1]

Application

4-Chlorotetrahydropyran was used in the synthesis of 3-chlorogutaric acid.[1]

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

114.8 °F - closed cup

flash_point_c

46 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses from 4-chlorotetrahydropyran
Stapp PR and Drake CA.
The Journal of Organic Chemistry, 36(4), 522-525 (1971)
Ursula Karges et al.
The Science of the total environment, 711, 134783-134783 (2019-12-11)
1,4-Dioxane, a cyclic ether that has been classified as a class 2B carcinogen by the US-EPA, is a substance of growing environmental concern because of its abundant occurrence in surface waters worldwide. Its high polarity and low biodegradability hamper its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service