Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330620

Sigma-Aldrich

4-Methyl-1-cyclohexanecarboxylic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H10CO2H
CAS Number:
Molecular Weight:
142.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

liquid

refractive index

n20/D 1.4598 (lit.)

bp

134-136 °C/15 mmHg (lit.)

density

1.005 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

CC1CCC(CC1)C(O)=O

InChI

1S/C8H14O2/c1-6-2-4-7(5-3-6)8(9)10/h6-7H,2-5H2,1H3,(H,9,10)

InChI key

QTDXSEZXAPHVBI-UHFFFAOYSA-N

Application

4-Methyl-1-cyclohexanecarboxylic acid was used to investigate the palladium core-porous silica shell-nanoparticles catalyzed hydrogenation of 4-carboxybenzaldehyde (4-CBA) to p-toluic acid.[1] It was also used in the synthesis of substituted cyclohexyl carbonyl chlorides.[2]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

>235.4 °F - closed cup

flash_point_c

> 113 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium core-porous silica shell-nanoparticles for catalyzing the hydrogenation of 4-carboxybenzaldehyde.
Li K-T, et al.
Catalysis Communications, 9(13), 2257-2260 (2008)
Synthesis and evaluation of mefway analogs as ligands for serotonin 5HT1A receptors.
Thio JP, et al.
Medicinal Chemistry Research, 1-7 null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service