Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

328642

Sigma-Aldrich

Borohydride, polymer-supported

support is Amberlyst® IRA-400, Macroporous, 20-50 mesh, extent of labeling: ~2.5 mmol/g loading

Synonym(s):

Borohydride on Amberlite IRA-400, (Polystyrylmethyl)trimethylammonium borohydride

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

beads

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: carbonyl reactive

extent of labeling

~2.5 mmol/g loading

particle size

20-50 mesh

SMILES string

[H][B-]([H])([H])[H].C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.C[N+](C)(C)Cc3ccccc3

InChI

1S/C4H12N.BH4/c1-5(2,3)4;/h1-4H3;1H4/q+1;-1

Inchi Key

LSSMEJCYZMCNIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for selective reduction of α,β-unsaturated aldehydes and ketones to allylic alcohols.[1]

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoon, N.M.
Tetrahedron Letters, 24, 5367-5367 (1983)
A. Nag et al.
Synthetic Communications, 17, 1007-1007 (1987)
Sande, A.R.
Tetrahedron Letters, 25, 3501-3501 (1984)
Gibson, H.W.;Bailey, J.
Journal of the Chemical Society, 815-815 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service