Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

326089

Sigma-Aldrich

Ammonium hexachloroiridate(IV)

98%

Synonym(s):

Iridium(IV)-ammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2IrCl6
CAS Number:
Molecular Weight:
441.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

assay

98%

form

powder

density

2.86 g/mL at 25 °C (lit.)

SMILES string

[H][N+]([H])([H])[H].[H][N+]([H])([H])[H].Cl[Ir--](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.Ir.2H3N/h6*1H;;2*1H3/q;;;;;;+4;;/p-4

InChI key

LWNOUTCTZQNGEN-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Duarte et al.
Nucleic acids research, 27(2), 496-502 (1998-12-24)
Oxidative damage to DNA bases commonly resultsin the formation of oxidized purines, particularly 7,8-dihydro-8-oxoguanine (8-oxoG) and 7,8-dihydro-8-oxoadenine (8-oxoA), the former being a well-known mutagenic lesion. Since 8-oxoG is readily subject to further oxidation compared with normal bases, the insertion of
G M Kolodny et al.
Cancer research, 43(7), 3101-3103 (1983-07-01)
Sodium hexachloroiridate injected 1 day after i.p. injection of 10(6) mouse ovarian tumor cells prevents the appearance of ascitic tumors in mice. Mice given injections of tumor cells all die at 20 to 30 days after tumor injection. Mice treated
Nootan Bhattarai et al.
Inorganic chemistry, 51(24), 13303-13311 (2012-11-29)
The aqueous oxidations of glutathione (GSH) by [IrCl(6)](2-), [Fe(bpy)(2)(CN)(2)](+), and [Fe(bpy)(CN)(4)](-) are described. All three reactions are highly susceptible to catalysis by traces of copper ions, but this catalysis can be fully suppressed with suitable chelating agents. The direct oxidation
Salah Boussaad et al.
Journal of the American Chemical Society, 130(12), 3780-3787 (2008-03-07)
In an effort to develop sensitive nanoscale devices for chemical and biological sensing, we have examined, using liquid gating, the conductance of semiconducting single-walled carbon nanotube-based field-effect transistors (SWCNT-FETs) in the presence of redox mediators. As examples, redox couples K3Fe(CN)6/K4Fe(CN)6
Na Song et al.
Inorganic chemistry, 47(24), 11458-11460 (2008-11-14)
One-electron oxidation of phenol, 2,4,6-trimethylphenol, and 2,6-dimethylphenol by [IrCl(6)](2-) in aqueous solution has a simple pH dependence, indicating slow bimolecular oxidation of ArOH and faster oxidation of ArO(-). H/D kinetic isotope effects as large as 3.5 for oxidation of ArOH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service