Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

324914

Sigma-Aldrich

Diethyl 2-(p-tolyl)malonate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH(CO2C2H5)2
CAS Number:
Molecular Weight:
250.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.492 (lit.)

bp

124-125 °C/1 mmHg (lit.)

density

1.054 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)C(C(=O)OCC)c1ccc(C)cc1

InChI

1S/C14H18O4/c1-4-17-13(15)12(14(16)18-5-2)11-8-6-10(3)7-9-11/h6-9,12H,4-5H2,1-3H3

InChI key

WGBSHZUHSDGHLV-UHFFFAOYSA-N

Application

Diethyl 2-(p-tolyl)malonate may be used in chemical synthesis.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service