Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

324442

Sigma-Aldrich

O2,2′-Cyclouridine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N2O5
CAS Number:
Molecular Weight:
226.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

optical activity

[α]19/D −21°, c = 4 in H2O

mp

248-251 °C (lit.)

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H]2[C@@H](OC3=NC(=O)C=C[N@@H]23)[C@@H]1O

InChI

1S/C9H10N2O5/c12-3-4-6(14)7-8(15-4)11-2-1-5(13)10-9(11)16-7/h1-2,4,6-8,12,14H,3H2/t4-,6-,7+,8-/m1/s1

InChI key

UUGITDASWNOAGG-CCXZUQQUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Research tool for antiviral and anticancer studies.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vladimir I Timofeev et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 63(Pt 10), 852-854 (2007-10-03)
Uridine phosphorylase (UPh; EC 2.4.2.3) is a member of the pyrimidine nucleoside phosphorylase family of enzymes which catalyzes the phosphorolytic cleavage of the C-N glycoside bond of uridine, with the formation of ribose 1-phosphate and uracil. This enzyme has been
Christopher S Theile et al.
Chemical communications (Cambridge, England), 48(45), 5587-5589 (2012-04-26)
Here we present new routes for the efficient syntheses of 6,5'-(S)- and 6,5'-(R)-cyclouridine. The syntheses utilize readily accessible uridine as a starting material. This route to the R diastereomer is significantly more efficient than previous synthetic efforts, allowing us to
A K Drabikowska et al.
Biochemical pharmacology, 36(23), 4125-4128 (1987-12-01)
Two series of 5-substituted uracil N(1)-acyclonucleosides, each with a different acyclic chain, were examined as inhibitors of uridine phosphorylase from rat intestinal mucosa, and several against the enzyme from Ehrlich ascites cells. In addition, several 5-substituted analogues of 2,2'-anhydrouridine were
Ying Fu et al.
Acta crystallographica. Section C, Crystal structure communications, 69(Pt 3), 282-284 (2013-03-06)
2,2'-Anhydro-1-(3',5'-di-O-acetyl-β-D-arabinofuranosyl)uracil, C13H14N2O7, was obtained by refluxing 2',3'-O-(methoxymethylene)uridine in acetic anhydride. The structure exhibits a nearly perfect C4'-endo ((4)E) conformation. The best four-atom plane of the five-membered furanose ring is O-C-C-C, involving the C atoms of the fused five-membered oxazolidine ring
Aldrichimica Acta, 20, 52-52 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service