Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

320668

Millipore

Petroleum ether

PRA grade

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12190000
grade:
PRA grade
bp:
30-60 °C (lit.)
vapor pressure:
24.47 psi ( 55 °C)
25.8 psi ( 55 °C)
7.51 psi ( 20 °C)
7.99 psi ( 20 °C)

grade

PRA grade

vapor density

2.5 (vs air)

vapor pressure

24.47 psi ( 55 °C)
25.8 psi ( 55 °C)
7.51 psi ( 20 °C)
7.99 psi ( 20 °C)

form

liquid

autoignition temp.

475 °F

expl. lim.

8 %

impurities

≤0.01% water
water

evapn. residue

<0.0001%

halogenated residue (GC/ECD)

<10 ng/L (as heptachlor epoxide)

refractive index

n20/D 1.363 (lit.)

bp

30-60 °C (lit.)

density

0.64 g/mL at 25 °C (lit.)

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

application

Suitable for pesticide residue analysis

pictograms

FlameHealth hazard

signalword

Danger

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

-56.2 °F - closed cup

flash_point_c

-49 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I P Dick et al.
Human & experimental toxicology, 16(11), 645-651 (1998-01-14)
1. The absorption and distribution of lindane through skin was examined using human volunteers. Two different preparations were employed, one with acetone as the vehicle and the other, a commercial product, consisting primarily of white spirit as the solvent base.
S C Bondy et al.
Archives of toxicology, 69(6), 410-414 (1995-01-01)
Levels of glutathione and activity of glutamine synthetase were assayed in organs of rats following inhalation of a heterogeneous solvent mixture containing both aliphatic and aromatic hydrocarbons. This mixture was administered for 3 weeks (6 h daily) at two levels
Catherine F Poh et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(22), 6716-6722 (2006-11-24)
Genetically altered cells could become widespread across the epithelium of patients with oral cancer, often in clinically and histologically normal tissue, and contribute to recurrent disease. Molecular approaches have begun to yield information on cancer/risk fields; tissue optics could further
H R Lam et al.
Neurotoxicology and teratology, 23(6), 603-608 (2002-01-17)
Previously, inhalation exposure to different types of white spirit (i.e. complex mixtures of aliphatic, aromatic, alkyl aromatic, and naphthenic hydrocarbons) has been shown to induce neurochemical effects in rat brains. Especially, the serotonergic system was involved at the global, regional
The use of a surgical glove in the prevention of tourniquet-associated chemical burns.
P J Tomlinson et al.
Annals of the Royal College of Surgeons of England, 90(3), 255-255 (2008-04-26)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service