Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

320455

Sigma-Aldrich

Phosphorus(V) oxychloride

ReagentPlus®, 99%

Synonym(s):

Phosphorus(V) oxide chloride, Phosphoryl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
POCl3
CAS Number:
Molecular Weight:
153.33
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

vapor density

5.3 (vs air)

vapor pressure

104 mmHg ( 50 °C)
28 mmHg ( 20 °C)

product line

ReagentPlus®

assay

99%

form

liquid

bp

105.8 °C (lit.)

mp

1.25 °C (lit.)

density

1.645 g/mL at 25 °C (lit.)

SMILES string

ClP(Cl)(Cl)=O

InChI

1S/Cl3OP/c1-5(2,3)4

InChI key

XHXFXVLFKHQFAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT RE 1 Inhalation

supp_hazards

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D M Badgujar et al.
Journal of hazardous materials, 172(1), 276-279 (2009-08-12)
1,2-Bis(2,4,6-trinitrophenyl) hydrazine (3) is one of the precursors in the synthesis of an important energetic material viz., hexanitrazobenzene. The simple and convenient lab scale synthesis of title compound (3) was carried out by the condensation of picryl chloride (2) with
Changjiang Yu et al.
Angewandte Chemie (International ed. in English), 51(31), 7688-7691 (2012-06-26)
Three for one: Pyrrolyldipyrromethenes having different functional groups were efficiently synthesized from POCl(3)-promoted condensations between 5-chloro-2-formylpyrrole or isoindole derivatives and suitable pyrrole or indole fragments through a novel nucleophilic aromatic substitution of the initially formed protonated azafulvene rings.
Simple coupling reaction between amino acids and weakly nucleophilic heteroaromatic amines.
Gilles Quéléver et al.
Journal of combinatorial chemistry, 6(5), 695-698 (2004-09-14)
Patrick J O'Brien et al.
Biochemistry, 41(9), 3207-3225 (2002-02-28)
Escherichia coli alkaline phosphatase (AP) is the prototypical two metal ion catalyst with two divalent zinc ions bound approximately 4 A apart in the active site. Studies spanning half a century have elucidated many structural and mechanistic features of this
Bo Qin et al.
Chemistry, an Asian journal, 6(12), 3298-3305 (2011-07-23)
POCl(3)-mediated one-pot macrocyclization allows the highly selective formation of five-residue macrocycles that are rigidified by internally placed intramolecular hydrogen bonds. Mechanistic investigation by using tailored competition experiments and kinetic simulation provides a comprehensive model, supporting a chain-growth mechanism underlying the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service