Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

317942

Fomblin® Y

LVAC 16/6, average mol wt 2700

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3O[-CF(CF3)CF2O-]x(-CF2O-)yCF3
CAS Number:
MDL number:
UNSPSC Code:
41105101
NACRES:
NB.79

mol wt

average mol wt 2700

refractive index

n20/D 1.299

viscosity

160 cSt(20 °C)(lit.)

density

1.89 g/mL at 25 °C

SMILES string

OC(O)(F)F.OC(F)(F)C(O)(F)C(F)(F)F

Looking for similar products? Visit Product Comparison Guide

General description

Unique perfluoropolyether vacuum oil is biologically and chemically inert, resists water, and is insoluble in most organic solvents. FOMBLIN Y has good dielectric properties and is compatible with plastomers, elastomers, and most metals over a wide temperature range. Has low vapor pressure, volatility, and surface tension. Also has high specific gravity, stability, and thermal resistance. 317985 and 317993 have very low dust and moisture content, making them ideal for high-purity applications. Thoroughly clean pump of any old fluid before adding Fomblin Y.

Legal Information

Fomblin is a registered trademark of Solvay Solexis S.P.A.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

>235.4 °F - closed cup

flash_point_c

> 113.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service