317462
Lithium aluminum hydride solution
0.5 M in ethylene glycol dimethyl ether
Synonym(s):
LAH, Lithium alanate, Lithium tetrahydroaluminate
About This Item
Recommended Products
reaction suitability
reagent type: reductant
concentration
0.5 M in ethylene glycol dimethyl ether
density
0.838 g/mL at 25 °C
SMILES string
[Li+].[AlH4-]
InChI
1S/Al.Li.4H/q-1;+1;;;;
InChI key
OCZDCIYGECBNKL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- The preparation of thermoplastic polyester polyamides from oleic acid
- Lithium-polymer batteries
- Hydrodefluorination of gem-difluoromethylene derivatives
- Asymmetric aldol reactions
- Synthesis of Li-Al-N-H composites with hydrogen absorption / desorption properties
LAH is a powerful reducing agent for many different reduction reactions such as that of ketones to alcohols
signalword
Danger
Hazard Classifications
Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2
supp_hazards
Storage Class
4.3 - Hazardous materials which set free flammable gases upon contact with water
wgk_germany
WGK 1
flash_point_f
41.0 °F
flash_point_c
5 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service